Distribution tubes with frit for specific heavy solvents
Proveedor: LENZ, E. (Lenz Laborglas GmbH & CO.KG)
05433202 05433203 05433204
LENZ05433202EA 41.9 EUR
LENZ05433202 LENZ05433203 LENZ05433204
Distribution tubes with frit for specific heavy solvents
Aparatos de extracción Extractores Soxhlet y tipo Soxhlet
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
Distribution tubes with frit (porosity: 0), for specific heavy solvents.