Test tubes, standard ground joints, without stopper
Proveedor: LENZ, E. (Lenz Laborglas GmbH & CO.KG)
05700034 05700007 05700010 05700030 05700014 05700026 05700038 05700042
LENZ05700034EA 8.6 EUR
LENZ05700034 LENZ05700007 LENZ05700010 LENZ05700030 LENZ05700014 LENZ05700026 LENZ05700038 LENZ05700042
Test tubes, standard ground joints, without stopper
Tubos Tubos de ensayos
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With standard ground joint, without stoppers.