Matraces Erlenmeyer, boca ancha, con reborde
Proveedor: LENZ, E. (Lenz Laborglas GmbH & CO.KG)
07025022 07025028 07025037 07025046 07025049 07025052 07025058 07025070 07025073
LENZ07025022EA 109 EUR
LENZ07025022 LENZ07025028 LENZ07025037 LENZ07025046 LENZ07025049 LENZ07025052 LENZ07025058 LENZ07025070 LENZ07025073
Matraces Erlenmeyer, boca ancha, con reborde
Matraces Matraces Erlenmeyer
DURAN®, borosilicate glass 3,3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and labelling area.