Invertebrates prepared slides
Proveedor: LIEDER JOHANNES
2101E 2102C 2103D 2105E 2107C 2108C 2109D 2116E 2117E 2124D
LIED2101EEA 10.2 EUR
LIED2101E LIED2102C LIED2103D LIED2105E LIED2107C LIED2108C LIED2109D LIED2116E LIED2117E LIED2124D
Invertebrates prepared slides
Portaobjetos Diapositivas preparadas
This set of invertebrates includes representative specimen of protozoa, sponges, coelenterata, worms, arthropoda, insects, molluscs, echinoderms and Acraniata.
Información de suministro: Supplied with accompanying brochure.