Culture Media, Ready To Use, In Plastic Tubes
Proveedor: LIOFILCHEM
26110P 26111P 26196P 26154P 26071P 26124P 26131P 26492P 26072P 26103P 24142P 26197P 26400P 26513P
LIOF26110PEA 237 EUR
LIOF26110P LIOF26111P LIOF26196P LIOF26154P LIOF26071P LIOF26124P LIOF26131P LIOF26492P LIOF26072P LIOF26103P LIOF24142P LIOF26197P LIOF26400P LIOF26513P
Culture Media, Ready To Use, In Plastic Tubes
Medios Medios preparados
Microbiology culture media in plastic tubes are utilised for cultivating, storing and observing microorganisms in various microbiological culture applications. Plastic tubes are increasingly being used for culture media instead of glass tubes, offering advantages compared to traditional glass tubes.
  • Reduced weight and lower CO₂ emissions: Plastic tubes are lighter than glass, resulting in lower transportation costs and a reduced carbon footprint
  • Enhanced safety: Plastic tubes are less likely to break during handling and transport, improving laboratory safety
  • Shelf life: Two years for most of the products (One year for the others)