Dehydrated Culture Media, Columbia Agar Base
Proveedor: LIOFILCHEM
6100135
LIOF6100135EA 796 EUR
LIOF6100135
Dehydrated Culture Media, Columbia Agar Base
Medios Medios deshidratados para microbiología
Columbia agar base, enriched with sterile sheep blood (5%), is suitable for isolation and growth of fastidious microorganisms such as streptococci, staphylococci, pneumococci and listeriae from clinical samples.