Agarose, I.D.NA™
Proveedor: LONZA
50170
LONZ50170EA 1080 EUR
LONZ50170
Agarose, I.D.NA™
Geles de electroforesis Agarosa y acrilamida
I.D.NA™ Agarose is a high gel strength, low electroendosmosis agarose developed for consistent resolution of DNA in DNA identity testing.
  • Free from DNase and RNase
  • Easy to handle gels that withstand the rigours of DNA identity tests
I.D.NA™ Agarose is certified under DNA identity testing conditions to exhibit clear DNA banding patterns necessary to discern VNTRs, HVRs, RFLPs, and DNA size standards. This high performance agarose displays high DNA transfer efficiency upon blotting and the lot to lot consistency required for DNA profiling tests.