Test Kits, molybdate (kits and Testpaks), CHECKIT®
Proveedor: Lovibond
147290 147295 147795 147291
LOVI147290EA 83.4 EUR
LOVI147290 LOVI147295 LOVI147795 LOVI147291
Test Kits, molybdate (kits and Testpaks), CHECKIT®
Kits para test medioambiente
CHECKIT® Comparator is a compact and handy colorimetric unit which is suitable for both mobile and static analysis work. Supplied with a generous number of different colour scales, it provides the basis for a comprehensive, easy-to use colorimetric analysis system.
  • Enables the use of large path lengths
  • The mirror optics makes use of the view through the entire length of the cell
  • Easy operation and exact reagent dosing
  • Tablet reagents have minimum guaranteed shelf life of 5/10 years
Testpak is a simple and cost-effective means of extending the use of an existing CHECKIT®Comparator instrument to a new test parameter.
Información de suministro: Test kit includes CHECKIT® discs, cells, stirring rod and AQUALYTIC® tablet reagents (for 30 tests) for the desired test supplied in a sturdy plastic case. Each Testpak contains the required CHECKIT® disc, tablet reagents (for 30 tests), two cells, stirring rod and detailed instructions for the desired test.