Florisil®, BAKER ANALYZED® 60 - 100 mesh Chromatography, J.T.Baker®

Proveedor: Avantor

Sinónimos: Ácido silícico, sal de magnesio, activado, Ácido silícico, sal de magnesio, Florisil

M368-07EA 257.73 EUR
M368-07
Florisil®, BAKER ANALYZED® 60 - 100 mesh Chromatography, J.T.Baker®
Silicato de magnesio

Selective adsorbent for gas chromatography. Contains activated magnesium silicate.

Formula: MgO₃Si
MW: 100.38 g/mol
Densidad: 2.51 Kg/L
Número MDL: MFCD00078249
Núm. CAS: 1343-88-0
EINECS: 215-681-1

Order Now

Resultados test especificaciones

For Laboratory,Research,or Manufacturing Use = 10 %
Mesh - On U.S. No. 60 Sieve ≤10 %
Mesh - Thru U.S. No. 100 Sieve ≤10 %
Appearance (fine, white crystalline powder). However, due to the high temperature involved in the activation process, some small
charred particles may be visible within the product.
Florisil® - Registered Trademark of Floridin Company


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR