Repromer Pb HPLC Columns
Proveedor: DR MAISCH
RM9.PB.S1078
MAISRM9.PB.S1078EA 828 EUR
MAISRM9.PB.S1078
Repromer Pb HPLC Columns
Columnas de cromatografía
For sugars, alcohols and organic acids: (polymers elute in front of monomers).
Order Now
ESPECIFICACIONES
Environmentally Preferable
Rango de pH 2 - 10