Crimp caps for crimp neck vials, N 20
Proveedor: MACHEREY-NAGEL
70233 70234 70242 70267 702928 702804 70234.8 70234.9 70235 70266 702805 702927 702928.9 702929 702799 702775 702094 702835 702808 70236.1 702773 702093 702128 702129 702834 702071 702112 702145 70234.10 702829 702833 702837 702838 702839
MANA70233EA 56 EUR
MANA70233 MANA70234 218-1272 MANA70267 218-1271 MANA702804 MANA70234.8 MANA70234.9 MANA70235 MANA70266 MANA702805 MANA702927 MANA702928.9 MANA702929 MANA702799 MANA702775 MANA702094 MANA702835 MANA702808 MANA70236.1 MANA702773 MANA702093 MANA702128 MANA702129 MANA702834 MANA702071 MANA702112 MANA702145 MANA70234.10 MANA702829 MANA702833 MANA702837 MANA702838 MANA702839
Crimp caps for crimp neck vials, N 20
Tapas Tapones para viales de cromatografía
Broad range of different crimp caps with mostly assembled septa, stoppers and PE caps are available for the crimp neck vials.
  • Center hole caps
  • Aluminium pressure release caps with special perforation for burst protection
  • Center tear off caps and complete tear off caps
  • Bi-metal crimp caps and magnetic crimp caps for autosamplers with magnetic transportation system
  • Silicone / PTFE septa (required above 120 °C) and Butyl / PTFE septa in different versions