Colorimetric rapid tests, chlorine, VISOCOLOR®
Proveedor: MACHEREY-NAGEL
920015 920115 931015 931090 931215 931290 935019 931016 931216 931217 931219 931035 936220.1 931235
MANA920015EA 119 EUR
MANA920015 MANA920115 MANA931015 MANA931090 MANA931215 MANA931290 MANA935019 MANA931016 MANA931216 MANA931217 MANA931219 MANA931035 MANA936220.1 MANA931235
Colorimetric rapid tests, chlorine, VISOCOLOR®
Kits para test medioambiente
The VISOCOLOR® testkits are devided into VISOCOLOR® alpha, ECO and HE. Every product range offering its specific strenght to provide a tailored test for each and every user and application.
  • Portable
  • Environmentally friendly
  • Convenient handling
  • Application-oriented measuring ranges
The addition of chlorine to swimming pools, water reservoirs and water mains is an approved procedure to rid the water of germs. With the correct dose harmful microorganisms are destroyed, many impurities removed and the growth of algae is prevented. However it is imperative that the chlorine content is regularly checked, since excessive chlorine not only impairs the smell and taste of the water but can be hazardous.
One distinguishes between free chlorine and bound chlorine (chloroamines); the sum of both is called total chlorine.