Colorimetric rapid tests, pH, VISOCOLOR®
Proveedor: MACHEREY-NAGEL
920074 920174 931066 931266 935075 931270
MANA920074EA 117 EUR
MANA920074 MANA920174 MANA931066 MANA931266 MANA935075 MANA931270
Colorimetric rapid tests, pH, VISOCOLOR®
Kits para test medioambiente
The VISOCOLOR® testkits are devided into VISOCOLOR® alpha, ECO and HE. Every product range offering its specific strenght to provide a tailored test for each and every user and application.
  • Portable
  • Environmentally friendly
  • Convenient handling
  • Application-oriented measuring ranges
The pH value indicates whether a water reacts acidic, alkaline or neutral. It is determined by the concentration of hydrogen ions. All biological processes in water are tied to specific pH ranges. For municipal and industrial applications, too, the control of specific pH limits is important since e. g. the efficiency of sewage plants or the corrosive action of tap water on the pipes depend in the pH value. Contrary to pH indicator papers VISOCOLOR® pH test kits can also be used to determine accurate pH values in unbuffered sample solutions.