NANOCOLOR® Photometric Tube Test Kits for COD

Proveedor: MACHEREY-NAGEL

985033 985028 963026 985023 985022 985012 985030 985011 985026 985027 985029 963029 985036 985038 985020
MANA985033EA 85.7 EUR
MANA985033 MANA985028 MANA963026 MANA985023 MANA985022 MANA985012 MANA985030 MANA985011 MANA985026 MANA985027 MANA985029 MANA963029 MANA985036 MANA985038 MANA985020
NANOCOLOR® Photometric Tube Test Kits for COD
Kits para test medioambiente

Photometric test kits for the determination of COD in water samples.


  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
  • Precisely pre-dosed reagents with constant high quality from batch to batch
  • Coloured pictogram instructions and easy handling of the tubes
  • 16 mm (OD) tubes for easy pipetting


The test kits are evaluated in 16 mm (OD) tubes. They are preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.


Sea water analysis: No

Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II



Información de suministro: Rugged box with 20 test tubes.

Order Now

ESPECIFICACIONES

Modelo MANA985033 MANA985028 MANA963026 MANA985023 MANA985022 MANA985012 MANA985030 MANA985011 MANA985026 MANA985027 MANA985029 MANA963029 MANA985036 MANA985038 MANA985020
Environmentally Preferable
Platform NANOCOLOR® tube tests NANOCOLOR® tube tests NANOCOLOR® tube tests NANOCOLOR® tube tests NANOCOLOR® tube tests NANOCOLOR® tube tests NANOCOLOR® tube tests NANOCOLOR® tube tests NANOCOLOR® tube tests NANOCOLOR® tube tests NANOCOLOR® tube tests NANOCOLOR® tube tests NANOCOLOR® tube tests NANOCOLOR® tube tests NANOCOLOR® tube tests
Temperatura de almacenamiento 15…25 °C 15…25 °C 2…8 °C 15…25 °C 2…8 °C 15…25 °C 15…25 °C 15…25 °C 15…25 °C 2…8 °C 15…25 °C 15…25 °C 15…25 °C 15…25 °C 2…8 °C
Periodo de validez (meses) 12 12 12 12 12 12 12 12 12 12 12 12 12 12 12
Peso 425 g (including packaging) 446 g (including packaging) 720.2 g (including packaging) 515.4 g (including packaging) 374 g (including packaging) 554 g (including packaging) 389.4 g (including packaging) 479 g (including packaging) 376 g (including packaging) 389.3 g (including packaging) 372.48 g (including packaging) 382 g (including packaging) 394 g (including packaging) 386 g (including packaging) 422.6 g (including packaging)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR