NANOCOLOR® Photometric Tube Test Kit for Fluoride
Proveedor: MACHEREY-NAGEL
985040
MANA985040EA 75.3 EUR
MANA985040
NANOCOLOR® Photometric Tube Test Kit for Fluoride
Kits para test medioambiente
Photometric test kit for the determination of fluoride in water samples.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
  • Precisely pre-dosed reagents with constant high quality from batch to batch
  • Coloured pictogram instructions and easy handling of the tubes
  • 16 mm (OD) tubes for easy pipetting
The test kit is evaluated in 16 mm (OD) tubes. It is preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Información de suministro: Rugged box with 20 test tubes.