Photometric test kits Robot tests
Proveedor: MACHEREY-NAGEL
985681 985603 985605 985607 985619 985621 985622 985626 985629 985630 985636 985638 985657 985664 985665 985668 985673 985674 985676 985680 985683 985688 985669
MANA985681EA 95.9 EUR
MANA985681 MANA985603 MANA985605 MANA985607 MANA985619 MANA985621 MANA985622 MANA985626 MANA985629 MANA985630 MANA985636 MANA985638 MANA985657 MANA985664 MANA985665 MANA985668 MANA985673 MANA985674 MANA985676 MANA985680 MANA985683 MANA985688 MANA985669
Photometric test kits Robot tests
Kits para test medioambiente
Robot SP2000series is a robotic analyser for the fully automated water analysis in the laboratory.
  • Automated water analysis with NANOCOLOR®
  • Fully automatic photometric water analysis including sample preparation and digestion
  • Analysis of all important waste water parameters
Robotic water analyser processes a variety of NANOCOLOR® tests and automates all the necessary handling steps of the photometric test kit procedure such as sample pipetting, (de)-capping, addition of reagents, mixing, heating, cooling and measurement. The flexibility and versatility of the test kit platform is for example reflected in the possibility to process from 24 to 336 test tubes at once by using different test tube racks.
The instrument can be configured for the analysis of one test kit application or for multiple test kit applications per sequence such as combinations of COD, total phosphate, total nitrogen and other parameters.