Chemical resistant gloves, Ultranitril 491
Proveedor: MAPA
34491266 34491267 34491268 34491260 34491269
MAPA34491266EA 270 EUR
MAPA34491266 MAPA34491267 MAPA34491268 MAPA34491260 MAPA34491269
Chemical resistant gloves, Ultranitril 491
Guantes
These anatomically designed gloves with flock lining provide good mechanical resistance and long-lasting chemical protection.
  • Dexterity and comfort thanks to the anatomical shape and the quality of the flock lining
  • Longer service life: Excellent mechanical resistance (abrasion, perforation)
  • Embossed texture for improved grip
Typical applications include: Automotive/mechanical industry, treating/degreasing metals with solvents, machining parts using cutting oil, maintenance, timber treatment and finishing, application of pesticides, cleaning ofMore Product Information
Order Now
ESPECIFICACIONES
Modelo Ultranitril 491
Environmentally Preferable
Ambidiestro/Específico para cada mano Específico para cada mano
Desechable / Reutilizable Reutilizable
Material del revestimiento Algodón
Acabado Embossed
Tipo de puño Turned & lined
Normativa EN EN 388:2016 (3101X), EN ISO 374-1 Type A (AJKOPT), EN ISO 374-5 VIRUS