Cut resistant gloves, KryTech 582
Proveedor: MAPA
34582037 34582030 34582031 34582038 34582039
MAPA34582037EA 26.5 EUR
MAPA34582037 MAPA34582030 MAPA34582031 MAPA34582038 MAPA34582039
Cut resistant gloves, KryTech 582
Guantes
These seamless HDPE liners with nitrile GRIP & PROOF coating are liquidproof and provide a good grip of oily parts.
  • Optimal cut resistance: level 5
  • Excellent resistance to abrasion
  • Visible indicator (blue textile) to easily identify the cut-performance level in the workplace
  • Performances of the gloves guaranteed after 5 cleaning cycles in washing machine
  • Non-slip grip
Ideal for metal working, the assembly of thin metal sheets, metal rolling and stamping, handling and sorting sharp parts, finishing work as well as for handling glass sheets.