Cold resistant gloves, TempIce 700
Proveedor: MAPA
34700017 34700018 34700019 34700010
MAPA34700017EA 163 EUR
MAPA34700017 MAPA34700018 MAPA34700019 MAPA34700010
Cold resistant gloves, TempIce 700
Guantes
These knitted gloves with insulating lining and Nitrile GRIP & PROOF coating on palm and fingertips provide protection in cold environment down to −10 °C while maintaining comfort, dexterity and flexibility.
  • Five times as durable as comparable standard gloves on the market
  • Excellent grip when handling wet parts
  • Stay dry when working in wet conditions
  • Washable (for five cleaning cycles)
  • Suitable for frozen, dry and some fatty foods use according to the European Directive CEE 1935/2004
Typical applications include: Order preparation, maintenance inMore Product Information