Slide folder, 'Munich'
Proveedor: MARIENFELD
6001313
MARI6001313EA 97.1 EUR
MARI6001313
Slide folder, 'Munich'
Portas
Cardboard base with three compartments (78×220 mm).
  • Each compartment can accomodate up to eight slides
  • Hinged cover with three windows (56×200 mm)