Safety scissors, SECUMAX 564
Proveedor: MARTOR ARGENTAX
564001.00
MARO564001.00EA 58 EUR
MARO564001.00
Safety scissors, SECUMAX 564
Tijeras
The SECUMAX 564 is characterised by its thumb opening and its extra-long edge, which also makes an ideal support surface. These safety scissors enable you to completely control your work.
  • Sharp, but not dangerous
  • Non-rusting stainless steel
  • Extra-long cutting edges
  • Robust, fiber-glass reinforced plastic handle
Highest safety guaranteed by the special sharpening of the cutting blades and the rounded tips.