Blasticidin S hydrochloride, en polvo, Corning®

Proveedor: Corning

30-100-RB
MDTC30-100-RBEA 236.72 EUR
MDTC30-100-RB
Blasticidin S hydrochloride, en polvo, Corning®
Blasticidin S hydrochloride

Blasticidin S HCl is produced by Streptomyces griseochromogenes and inhibits translation in both prokaryotes and eukaryotes, thus preventing the formation of peptide bonds.

Formula: C₁₇H₂₇ClN₈O₅
Temperatura de almacenaje: Congelador
Número MDL: MFCD02091640
Núm. CAS: 3513-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR