IPTG (Isopropil-β-D-tiogalactopiranósido) ≥99%, en polvo

Proveedor: Corning

Sinónimos: IPTG

46-102-RF
MDTC46-102-RFEA 55.9 EUR
MDTC46-102-RF
IPTG (Isopropil-β-D-tiogalactopiranósido) ≥99%, en polvo
IPTG (Isopropil-β-D-tiogalactopiranosida)

IPTG (isopropylthiogalactoside) is an inducer of the lac operon in bacteria, which is frequently used in cloning as a component of a recombinant plasmid.


When used with X-gal to detect lac gene activity, IPTG can be used for screening of clones containing a gene of interest in the lac operon.

Formula: C₉H₁₈O₅S
Pto. Ebullición: 402 °C (1013 hPa)
Pto. de fusión: 109,5…110.5 °C
Temperatura de almacenaje: Congelador
Número MDL: MFCD00063273
Núm. CAS: 367-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR