Cryopreservation bags, Corning®
Proveedor: Corning
91-200-89 91-200-90 91-200-88 91-200-91
MDTC91-200-89EA 71.45 EUR
MDTC91-200-89 MDTC91-200-90 MDTC91-200-88 MDTC91-200-91
Cryopreservation bags, Corning®
Bolsas Bolsas de medios
These cryogenic storage bags are designed for storage, preservation, and transfer of cells.
  • Unique bag film material remains flexible at ultra-low temperatures
A proprietary membrane port design offers thinner walls for increased flexibility, while an attached cap minimizes membrane exposure during freezing. Industry standard label pocket design ensures ease-of-use and traceability in labeling.
Port 1: Two thin-walled membrane ports.
Port 2: 4,5 L×0,153" Ext.Ø EVA tube, splits to two 5 L×0,118 Int.Ø×0,16" Ext.Ø PVC tubes with female Luer connectorsMore Product Information