Incubators, Generation 2012, IN/INplus (natural convection)
Proveedor: MEMMERT
IN110 IN110PLUS IN160 IN160PLUS IN260 IN260PLUS IN30 IN30PLUS IN450 IN450PLUS IN55 IN55PLUS IN75 IN750 IN750PLUS IN75PLUS IN160M IN160MPLUS IN450MPLUS IN55M IN750M IN750MPLUS IN260M IN260MPLUS IN55MPLUS IN110M IN30M IN30MPLUS IN75M IN75MPLUS IN110MPLUS IN450M
390-0644EA 2360 EUR
390-0644 390-0645 390-0646 390-0647 390-0648 390-0649 390-0650 390-0651 390-0652 390-0653 390-0654 390-0655 390-0656 390-0657 390-0658 390-0659 MEMMIN160M MEMMIN160MPLUS MEMMIN450MPLUS MEMMIN55M MEMMIN750M MEMMIN750MPLUS MEMMIN260M MEMMIN260MPLUS MEMMIN55MPLUS MEMMIN110M MEMMIN30M MEMMIN30MPLUS MEMMIN75M MEMMIN75MPLUS MEMMIN110MPLUS MEMMIN450M
Incubators, Generation 2012, IN/INplus (natural convection)
Incubadoras Incubadores estándar
The range has natural and forced convection models with a temperature range up to 80 °C. There is a choice of two different displays, SingleDISPLAY or TwinDISPLAY depending on the application.
  • SingleDISPLAY models, ideal for standard applications:
  • ControlCOCKPIT parameters: Temperature (Celsius or Fahrenheit), fan speed, exhaust air flap position, program time, time zones and daylight-saving time
  • One PT100 temperature sensor in a four wire circuit and Ethernet interface for exporting the protocol log
  • Double over-temperature protection including visual