Masterflex® Variable Area Flowmeter Multitube Frames, Avantor®
Proveedor: Avantor Fluid Handling
03214-28 03214-30 03214-36 03214-46 03214-52 03214-56 03214-64 03214-66 03214-68 03214-74 03214-76 03214-78 03214-80 03215-28 03215-46 03215-64 03215-74 03215-80
MFLX03214-10EA 154 EUR
MFLX03214-10 MFLX03214-12 MFLX03214-16 MFLX03214-18 MFLX03214-50 MFLX03214-62 MFLX03215-10 MFLX03215-12 MFLX03215-16 MFLX03215-18 MFLX03215-30 MFLX03215-34 MFLX03215-36 MFLX03215-50 MFLX03215-52 MFLX03215-56 MFLX03215-68 MFLX03215-76 MFLX03215-78 MFLX03214-28 MFLX03214-30 MFLX03214-36 MFLX03214-46 MFLX03214-52 MFLX03214-56 MFLX03214-64 MFLX03214-66 MFLX03214-68 MFLX03214-74 MFLX03214-76 MFLX03214-78 MFLX03214-80 MFLX03215-28 MFLX03215-46 MFLX03215-64 MFLX03215-74 MFLX03215-80
Masterflex® Variable Area Flowmeter Multitube Frames, Avantor®
Medidores de flujo
Create your own flowmeter systems for complete flexibility.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Use with media for laboratory and industrial applications
  • Required system components for flowmeter systems would be frame, flowtube(s), valve cartridge, and tripod—order each component separately
  • Choose from aluminum or stainless