Masterflex® Transfer Tubing, Perfluoroalkoxy (PFA), Avantor®
Proveedor: Avantor Fluid Handling
06371-07 06371-08 06371-22 06371-23
MFLX06371-00EA 56.8 EUR
MFLX06371-00 MFLX06371-01 MFLX06371-02 MFLX06371-03 MFLX06371-04 MFLX06371-05 MFLX06371-06 MFLX06371-21 MFLX06371-24 MFLX06371-25 MFLX06371-26 MFLX06371-28 MFLX06371-29 MFLX06375-01 MFLX06375-02 MFLX06375-04 MFLX06375-05 MFLX06375-07 MFLX06375-08 MFLX06371-07 MFLX06371-08 MFLX06371-22 MFLX06371-23
Masterflex® Transfer Tubing, Perfluoroalkoxy (PFA), Avantor®
Tuberías Tubo de transferencia
Stands up to highly corrosive materials.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. For the transfer of high-temperature or highly corrosive materials. Also good for high-purity work.
Characteristics:More Product Information