Masterflex® Chemically Inert Metering Valves, Avantor®
Proveedor: Avantor Fluid Handling
06394-00 06394-02 06394-04 06394-16
MFLX06394-00EA 136 EUR
MFLX06394-00 MFLX06394-02 MFLX06394-04 MFLX06394-16
Masterflex® Chemically Inert Metering Valves, Avantor®
Válvulas Válvulas dosificadoras
Easily meter corrosive fluids or gases.
  • Just turn—eight turns fully open or close valv
Excellent for metering corrosive fluids and gases — all wetted parts are CTFE and PTFE. Eight turns will fully open or close valves. Maximum pressure is 75 psi and maximum temperature is 150 °F (93 °C); orifice size is 1/4".