Masterflex® L/S® Precision Pump Tubing, Tygon® E-Food, Avantor®
Proveedor: Avantor Fluid Handling
MFLX06418-13EA 83.9 EUR
MFLX06418-13 MFLX06418-14 MFLX06418-16 MFLX06418-17 MFLX06418-18 MFLX06418-25
Masterflex® L/S® Precision Pump Tubing, Tygon® E-Food, Avantor®
Tuberías Tubería de bomba peristáltica
Designed specifically for handling food products.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for