Masterflex® I/P® Precision Pump Tubing, C-Flex®, Avantor®
Proveedor: Avantor Fluid Handling
MFLX06424-26EA 212 EUR
MFLX06424-26 MFLX06424-73 MFLX06424-82
Masterflex® I/P® Precision Pump Tubing, C-Flex®, Avantor®
Tuberías Tubería de bomba peristáltica
Maintain flexibility at extreme temperatures.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory Approvals to match your unique application
  • Ultra smooth inner surface results in fewer extractables
Masterflex®More Product Information