Masterflex® I/P® Precision Pump Tubing, C-Flex® ULTRA, Avantor®
Proveedor: Avantor Fluid Handling
MFLX06434-73EA 229 EUR
MFLX06434-73 MFLX06434-82 MFLX06434-26 MFLX06437-73
Masterflex® I/P® Precision Pump Tubing, C-Flex® ULTRA, Avantor®
Tuberías Tubería de bomba peristáltica
Ultra performance for ultrapure applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory Approvals to match your unique application
  • Longest pumping life of any C-Flex® formulation
Masterflex® C-Flex®More Product Information