Single Open-End Absolute-Rated Cartridge Systems
Proveedor: Avantor Fluid Handling
06479-18 06479-34 06479-46 06479-62 06479-42
MFLX06479-18EA 272 EUR
MFLX06479-18 MFLX06479-34 MFLX06479-46 MFLX06479-62 MFLX06479-42
Single Open-End Absolute-Rated Cartridge Systems
Filtros Cartuchos de filtros
A collection to fit a wide range of critical applications.
  • Absolute-rated for use in many regulated processes
  • Meets a wide range of industry and government standards
These SOE cartridges have a 222 O-ring configuration to create a tight seal and prevent influent bypass. They are autoclavable to 250 °F (121 °C) and in-line sterilizable to 257 °F (125 °C) for up to 30-minute cycles; tolerant to an accumulated exposure of 10 hours. The filters may also be sanitized with compatible chemical agents. All cartridges are suitable to 180 °F (82 °C) at 10 psid.More Product Information