Standard-Wall PTFE Heat-Shrink Transfer Tubing
Proveedor: Avantor Fluid Handling
06851-22 06851-42
MFLX06851-22EA 96.7 EUR
MFLX06851-22 MFLX06851-42
Standard-Wall PTFE Heat-Shrink Transfer Tubing
Tuberías Tubo termorretráctil
Ideal for chemically inert applications.
  • High electrical and chemical resistance
  • Comes in 3-ft lengths
  • Opaque
Shrink ratio: 2:1
Recovery temperature: 621 °F (327 °C)
Characteristics: Opaque; high electrical and chemical resistance.