FMI Valveless Piston Pump Heads, Series Q
Proveedor: Avantor Fluid Handling
MFLX07104-53EA 1820 EUR
MFLX07104-53 MFLX07104-54 MFLX07104-56 MFLX07104-63 MFLX07104-64 MFLX07104-66
FMI Valveless Piston Pump Heads, Series Q
Bombas Cabezales de bomba Cabezas de bombas de pistón
No valves to clog or wear out.
  • Build your own pump systems using interchangeable heads, drives, and accessories
  • Pump fluids with viscosities up to 500 cp
  • Low dead volume
These FMI Series Q valveless piston pump heads give you options to handle a wide range of fluids at pressures up to 100 psi. The interchangeable pump head modules provide numerous flow and material combinations for optimal versatility. Choose from a combination of ceramic, PVDF, carbon, and stainless steel wetted materials for an inert and corrosion resistant fluid path, ideal forMore Product Information