Pump Motor Coupling Hardware and Accessories
Proveedor: Avantor Fluid Handling
MFLX07127-59EA 28.7 EUR
MFLX07127-59
Pump Motor Coupling Hardware and Accessories
Bombas Motores
Create your own motor mounting with the hardware below.
  • Optimize performance of your Pump Motor
Order one coupling with the same diameter as the motor driving shaft, one coupling with the same diameter as the driven shaft, and one spider (see accessories).