Masterflex® L/S® Easy-Load® Long-Shaft Pump Heads, Avantor®
Proveedor: Avantor Fluid Handling
MFLX07514-50EA 584 EUR
MFLX07514-50 MFLX07514-52 MFLX07514-60
Masterflex® L/S® Easy-Load® Long-Shaft Pump Heads, Avantor®
Bombas Bombas diseñadas a medida
Save time with fast tubing changes and reduced maintenance.
  • New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
  • Rotor is constructed of stainless steel (SS) for enhanced durability
This L/S® Easy-Load® II Pump Head, when combined with a Masterflex® L/S® drive or compatible system, is designed to provide a simple, easy-to-use peristaltic pump system. The pump head accepts several different tubing sizes for a wide range of flow rates.