Masterflex® L/S® Digital Miniflex® Pump Systems, Avantor®
Proveedor: Avantor Fluid Handling
MFLX07525-20-EUEA 2900 EUR
MFLX07525-20-EU MFLX07525-40-EU
Masterflex® L/S® Digital Miniflex® Pump Systems, Avantor®
Bombas Bombas peristálticas
Make low-flow pumping easy!
  • Flow range 0.004 to 315 ml/min—flow rate depends on drive rpm, pump head, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating mode
  • Remote control available using analog communication—voltage, current and contact closure
  • Integrated “open-head” sensor halts operation when pump head is opened
  • Pump head accepts continuous lengths of tubing for clean flow paths
  • Automatic tubing retention makes tube loading quick and easy
  • More than 20% smaller than competitive units—compact size and light weight