Masterflex® Universal Pressure Sensor Adapter Cable, Avantor®
Proveedor: Avantor Fluid Handling
MFLX07526-99EA 134 EUR
MFLX07526-99
Masterflex® Universal Pressure Sensor Adapter Cable, Avantor®
Bombas Bombas peristálticas
Adapts a variety of sensors to the MasterSense™ Sensor AUX input.
  • Designed for use with 0-20/4-20 mA or 0-5,0-10 V sensors
  • Includes 24VDC power out pins for loop powered circuits up to 50 mA
Connection cable allows for integration of variety of sensors into Masterflex® MasterSense™ pump systems. Reference MasterSense™ instruction manual for wiring diagrams and detail.