Masterflex® Strainer Tops for Low-Cost In-Line Strainer Systems, Avantor®
Proveedor: Avantor Fluid Handling
29595-01 29595-03 29595-05 29595-07 29595-09 29595-13 29595-15 29595-17 29595-11
MFLX29595-01EA 21.4 EUR
MFLX29595-01 MFLX29595-03 MFLX29595-05 MFLX29595-07 MFLX29595-09 MFLX29595-13 MFLX29595-15 MFLX29595-17 MFLX29595-11
Masterflex® Strainer Tops for Low-Cost In-Line Strainer Systems, Avantor®
Filtros Filtros
Easily customize strainer system to suit your application.
  • Choose from three strainer top options based on required filtration area
Build your own economical filtration system to filter out particulates from your fluids. Available in three classes: low-profile, intermediate, and large. Modify or change components of the same class to meet the demands of your application.
Low-Profile Strainers are ideal for small-scale low-volume filtration. Use in research applications, cleaning equipment, and laboratory equipment.
Intermediate Strainers feature a largeMore Product Information