Masterflex® Manifold Fittings, Female Luer Locks, Avantor®
Proveedor: Avantor Fluid Handling
MFLX30600-41EA 114 EUR
MFLX30600-41 MFLX30600-42 MFLX30600-43 MFLX30600-44
Masterflex® Manifold Fittings, Female Luer Locks, Avantor®
Válvulas Válvulas de bola
Regulate flow without any leaking.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs,