Masterflex® Gamma Irradiated Fittings, Sanitary Clamp to Barb with TPE Fused Gasket, Avantor®
Proveedor: Avantor Fluid Handling
MFLX31201-24EA 284 EUR
MFLX31201-24 MFLX31201-14 MFLX31201-16 MFLX31201-17 MFLX31201-18 MFLX31201-23 MFLX31201-25 MFLX31201-15 MFLX31201-19 MFLX31201-20
Masterflex® Gamma Irradiated Fittings, Sanitary Clamp to Barb with TPE Fused Gasket, Avantor®
Racores y conectores
Sanitary fitting with built-in gasket makes sanitary installation easier than ever.
  • Presterilized adapters cut down on lead time and are available for quick shipment
  • Sanitary gasket combined with a sanitary fitting adds another layer of safety to installation
  • Prevent contamination with zero user contact with the gasket to ensure sterility
  • Eliminate the risk of misalignment with the limited slip design
Allowing for easy assembly and disassembly of sanitary setups, these adapters are suitable for use connecting a Tri-Clamp® fitting to flexible tubingMore Product Information