Masterflex® Barbed Medium Flow Female Quick-Disconnect Fittings, Avantor®
Proveedor: Avantor Fluid Handling
MFLX40250-02EA 86.4 EUR
MFLX40250-02 MFLX40250-03 MFLX40250-10 MFLX40250-11 MFLX40250-17 MFLX40250-18 MFLX40250-25 MFLX40250-26 MFLX40250-32 MFLX40250-33 MFLX40250-40 MFLX40250-41 MFLX40250-47 MFLX40250-48 MFLX40250-55 MFLX40250-56
Masterflex® Barbed Medium Flow Female Quick-Disconnect Fittings, Avantor®
Racores y conectores
Mechanical innovations reduce risk of fluid cross-over and contamination. A mating connection is made with a Medium Flow Male fitting.
  • Aseptic connect and disconnect
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass through at lower line pressures in a smaller body. As the overall quick disconnect design has matured,More Product Information