Masterflex® 3/4" Sanitary Clamp Medium Flow Male Quick-Disconnect Fittings, Avantor®
Proveedor: Avantor Fluid Handling
MFLX40257-42EA 89.6 EUR
MFLX40257-42 MFLX40257-43 MFLX40257-49 MFLX40257-50 MFLX40257-56 MFLX40257-57 MFLX40257-63 MFLX40257-64 MFLX40257-70 MFLX40257-71 MFLX40257-77 MFLX40257-78 MFLX40257-79
Masterflex® 3/4" Sanitary Clamp Medium Flow Male Quick-Disconnect Fittings, Avantor®
Racores y conectores
Mechanical innovations reduce risk of fluid cross-over and contamination. A mating connection is made with a Medium Flow Female fitting.
  • Aseptic connect and disconnect
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass through at lower line pressures in a smaller body. As the overall quick disconnect design hasMore Product Information