Masterflex® Adapter Fittings, Sanitary Clamp to Male Thread, Straight, Avantor®
Proveedor: Avantor Fluid Handling
MFLX31200-03EA 151 EUR
MFLX31200-03 MFLX31805-59 MFLX31805-63 MFLX31805-71 MFLX40619-36 MFLX40619-39 MFLX40619-41 MFLX40619-47 MFLX40619-51
Masterflex® Adapter Fittings, Sanitary Clamp to Male Thread, Straight, Avantor®
Racores y conectores
Maintain sanitary conditions while quickly and easily connecting to a male NPT thread.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for