Masterflex® Adapter Fittings, Luer to Luer, Polycarbonate, Avantor®
Proveedor: Avantor Fluid Handling
45501-22
MFLX30526-10EA 30.1 EUR
MFLX30526-10 MFLX45501-28 MFLX45501-56 MFLX45501-82 MFLX45501-84 MFLX45504-56 MFLX45501-22 MFLX45501-64
Masterflex® Adapter Fittings, Luer to Luer, Polycarbonate, Avantor®
Racores y conectores
Easily transition to a luer fitting.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs,