Masterflex® High-Precision, High-Pressure Piston Pumps, Avantor®
Proveedor: Avantor Fluid Handling
MFLX74931-00EA 4090 EUR
MFLX74931-00 MFLX74931-01 MFLX74931-10 MFLX74931-12 MFLX74931-20 MFLX74931-45 MFLX74931-11 MFLX74931-30
Masterflex® High-Precision, High-Pressure Piston Pumps, Avantor®
Bombas Bombas de pistón
The right pump for your HPLC application.
  • Flow rates from 10 to 300 ml/min
  • Digital stepper motor ensures accuracy at low flow rates
  • Remotely control pump with a PC via RS-232 and micro USB
  • Self-flush feature washes piston with every stroke
  • Interactive keypad controls make programming at the pump
Single-Head Pumps precisely meter and dispense chemicals, perfect for small-scale laboratory fluid transfers and HPLC applications. Small footprint saves bench and fume hood space.
High-Flow Dual-Head Pumps are ideally suited to Preparative
Información deMore Product Information