Masterflex® Laboratory Software and Prepackaged Accessories, Avantor®
Proveedor: Avantor Fluid Handling
MFLX78002-92EA 66.4 EUR
MFLX78002-92
Masterflex® Laboratory Software and Prepackaged Accessories, Avantor®
Software Software de laboratorio
Stand-alone process operation. No need for a PC.
  • Optimize performance of your Masterflex pumps
The ProgEdit software enables the user to program individually definable dispensing and pumping sequences with a PC and to download the program to the memory of the pump drive.
ISMATEC's MCP Process dispensing pumps are ideal for safe dispensing and filling applications. Individual, application based dispensing and pumping sequences can be programmed directly into a PC (batch-programming) and subsequently downloaded (via the RS232 interface) into one of theMore Product Information