Masterflex® Ismatec® Reglo Digital Piston Pump Systems with MasterflexLive®, Avantor®
Proveedor: Avantor Fluid Handling
MFLX78018-64-EUEA 4880 EUR
MFLX78018-64-EU MFLX78018-65-EU MFLX78018-66-EU MFLX78018-63 MFLX78018-63-AU MFLX78018-63-CH MFLX78018-63-DK MFLX78018-63-EU MFLX78018-63-IN MFLX78018-63-IT MFLX78018-63-UK
Masterflex® Ismatec® Reglo Digital Piston Pump Systems with MasterflexLive®, Avantor®
Bombas Bombas de pistón
Achieve high pressure, accuracy, and repeatability—with an intuitive touch-screen interface.
  • MasterflexLive® is a secure, cloud-enabled platform for monitoring Masterflex pumps
  • Compact, benchtop pumps optimize ease of use, accuracy, and reliability
  • 5” (12.7 cm) capacitive TFT/RGB touch screen features crisp resolution and a wide viewing angle
  • Flexible operational control via screen or analog remote control
  • Continuous run, volume dispense, and time dispense modes—batch control and interval/off time in volume and time modes
  • Pump allows for saving and