Masterflex® Ismatec® Microbore Pump Tubing, Platinum-Cured Silicone, Avantor®
Proveedor: Avantor Fluid Handling
MFLX95590-26EA 295 EUR
MFLX95590-26 MFLX95590-30 MFLX95590-34 MFLX95590-42 MFLX95590-48
Masterflex® Ismatec® Microbore Pump Tubing, Platinum-Cured Silicone, Avantor®
Tuberías Tubería de bomba peristáltica
Smooth surface and low protein binding levels.
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
Use platinum-cured silicone tubing for high purity applications where outgassing of peroxide product is not allowed.
For custom tubing lengths andMore Product Information