Tygoprene® XL-60 Transfer Pump Tubing

Proveedor: Avantor Fluid Handling
95870-00 95870-02 95870-06 95870-08 95870-10 95870-12
MFLX95870-00EA 69.9 EUR
MFLX95870-00 MFLX95870-02 MFLX95870-06 MFLX95870-08 MFLX95870-10 MFLX95870-12
Tygoprene® XL-60 Transfer Pump Tubing
Tuberías Tubo de transferencia
Use in varying temperatures.

  • FDA compliant for food contact

Formulation: Tygoprene XL-60
Applications: Ink transfer, soap and detergent delivery, cold or hot beverage transfer and dispenseing, food processing, general chemical transfer and processing.
Characteristics: Low extractables, excellent resistance to ozone, oxygen, and sunlight. Resists a wide range of fluids, including acids and bases. Translucent.
Certification: FDA compliant. Tubing is NSF listed under standard 51 and is UL 94-HB flammability resistant.
Temperature range: — 87 to 250 °F ( — 66More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR